Tisk…

1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 95%

BLDPBD90456-250MGEA 285 CZK
BLDPBD90456-250MG BLDPBD90456-25G BLDPBD90456-1G BLDPBD90456-10G BLDPBD90456-100G BLDPBD90456-5G
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 95%
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₃H₂₀BrN₃O₂
MW: 330,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09831980
CAS číslo: 877399-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR