Tisk…

4-(Difluoromethoxy)benzenesulphonamide 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD91224-1GEA 1340 CZK
BLDPBD91224-1G BLDPBD91224-10G BLDPBD91224-5G BLDPBD91224-25G
4-(Difluoromethoxy)benzenesulphonamide 98%
4-(Difluoromethoxy)benzenesulphonamide

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₇F₂NO₃S
MW: 223,2000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03407973
CAS číslo: 874781-09-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR