4-(Difluoromethoxy)benzylamine 95%
|
|
BLDPBD91233-250MGEA
257
CZK
BLDPBD91233-250MG
BLDPBD91233-1G
BLDPBD91233-5G
4-(Difluoromethoxy)benzylamine 95%
4-(Difluoromethoxy)benzylamine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₈H₉F₂NO MW: 173,16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00236231 CAS číslo: 177842-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...