4-(Methylthio)benzonitrile 98%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD91436-5GEA
1090
CZK
BLDPBD91436-5G
BLDPBD91436-1G
BLDPBD91436-25G
4-(Methylthio)benzonitrile 98%
4-(Methylthio)benzonitrile
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₈H₇NS MW: 149,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010381 CAS číslo: 21382-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...