Tisk…

4-(Heptyloxy)phenylboronic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD91501-5GEA 969 CZK
BLDPBD91501-5G BLDPBD91501-10G BLDPBD91501-1G BLDPBD91501-250MG BLDPBD91501-100G BLDPBD91501-25G
4-(Heptyloxy)phenylboronic acid 97%
4-(Heptyloxy)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₃H₂₁BO₃
MW: 236,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039169
CAS číslo: 136370-19-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR