Tisk…

4,5-Dichloro-3(2H)-pyridazinone 97%

BLDPBD92532-1000GEA 13510 CZK
BLDPBD92532-1000G BLDPBD92532-500G BLDPBD92532-5G BLDPBD92532-25G BLDPBD92532-100G BLDPBD92532-10G
4,5-Dichloro-3(2H)-pyridazinone 97%
4,5-Dichloro-3(2H)-pyridazinone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₄H₂Cl₂N₂O
MW: 164,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051504
CAS číslo: 932-22-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR