Resodiacetophenone 97%
|
|
BLDPBD92726-25GEA
6100
CZK
BLDPBD92726-25G
BLDPBD92726-5G
BLDPBD92726-10G
BLDPBD92726-1G
Resodiacetophenone 97%
Resodiacetophenone
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₁₀H₁₀O₄ MW: 194,1840 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017655 CAS číslo: 2161-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...