Tisk…

4,6-Dichloro-2,5-dimethylpyrimidine 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD92771-5GEA 3250 CZK
BLDPBD92771-5G BLDPBD92771-1G BLDPBD92771-250MG
4,6-Dichloro-2,5-dimethylpyrimidine 96%
4,6-Dichloro-2,5-dimethylpyrimidine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₆Cl₂N₂
MW: 177,031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991674
CAS číslo: 1780-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR