Tisk…

4-(Morpholinomethyl)phenylboronic acid pinacol ester 95%

BLDPBD93248-25GEA 4340 CZK
BLDPBD93248-25G BLDPBD93248-10G BLDPBD93248-5G BLDPBD93248-1G BLDPBD93248-250MG
4-(Morpholinomethyl)phenylboronic acid pinacol ester 95%
4-(Morpholinomethyl)phenylboronic acid pinacol ester

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₇H₂₆BNO₃
MW: 303,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974052
CAS číslo: 364794-79-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR