Tisk…

4-Acetamidothiophenol 98%

BLDPBD93374-25GEA 3400 CZK
BLDPBD93374-25G BLDPBD93374-1G BLDPBD93374-5G
4-Acetamidothiophenol 98%
4-Acetamidothiophenol

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₉NOS
MW: 167,2282 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004848
CAS číslo: 1126-81-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR