Tisk…

4-Allyloxy-2-hydroxybenzophenone 98%

BLDPBD93498-10GEA 371 CZK
BLDPBD93498-10G
4-Allyloxy-2-hydroxybenzophenone 98%
4-Allyloxy-2-hydroxybenzophenone

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₆H₁₄O₃
MW: 254,2806 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191713
CAS číslo: 2549-87-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR