Tisk…

4'-Benzyloxy-2'-hydroxyacetophenone 98%

BLDPBD94454-250MGEA 314 CZK
BLDPBD94454-250MG BLDPBD94454-5G BLDPBD94454-100MG BLDPBD94454-25G BLDPBD94454-10G BLDPBD94454-100G BLDPBD94454-1G
4'-Benzyloxy-2'-hydroxyacetophenone 98%
4'-Benzyloxy-2'-hydroxyacetophenone

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₅H₁₄O₃
MW: 242,2699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054612
CAS číslo: 29682-12-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR