4-Bromo-1,8-naphthalic anhydride 96%
|
|
BLDPBD94659-500GEA
2660
CZK
BLDPBD94659-500G
BLDPBD94659-1000G
BLDPBD94659-5G
BLDPBD94659-10G
BLDPBD94659-25G
BLDPBD94659-100G
4-Bromo-1,8-naphthalic anhydride 96%
4-Bromo-1,8-naphthalic anhydride
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₁₂H₅BrO₃ MW: 277,0703 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006927 CAS číslo: 81-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...