Tisk…

4-Bromo-1-ethyl-1H-pyrazole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD94671-5GEA 1860 CZK
BLDPBD94671-5G BLDPBD94671-1G BLDPBD94671-10G BLDPBD94671-25G BLDPBD94671-100G
4-Bromo-1-ethyl-1H-pyrazole 97%
4-Bromo-1-ethyl-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₅H₇BrN₂
MW: 175,0265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235252
CAS číslo: 71229-85-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR