Tisk…

4-Bromo-2,3-difluorobenzoic acid 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD94753-500GEA 92800 CZK
BLDPBD94753-500G BLDPBD94753-5G BLDPBD94753-25G BLDPBD94753-100MG BLDPBD94753-0.5G BLDPBD94753-10G BLDPBD94753-100G BLDPBD94753-1G BLDPBD94753-250MG
4-Bromo-2,3-difluorobenzoic acid 96%
4-Bromo-2,3-difluorobenzoic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₃BrF₂O₂
MW: 236,9983 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368121
CAS číslo: 194804-91-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR