Tisk…

4-Bromo-2,5-xylidine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD94773-100GEA 6330 CZK
BLDPBD94773-100G BLDPBD94773-25G BLDPBD94773-5G BLDPBD94773-1G
4-Bromo-2,5-xylidine 98%
4-Bromo-2,5-xylidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₁₀BrN
MW: 200,0757 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780642
CAS číslo: 30273-40-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR