4-Bromo-4'-methoxybiphenyl 98%
|
|
BLDPBD95103-5GEA
2570
CZK
BLDPBD95103-5G
BLDPBD95103-1G
4-Bromo-4'-methoxybiphenyl 98%
4-Bromo-4'-methoxybiphenyl
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₁₃H₁₁BrO MW: 263,1298 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00671954 CAS číslo: 58743-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...