Tisk…

4-Bromo-6-methoxyquinoline 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD95143-1GEA 3540 CZK
BLDPBD95143-1G BLDPBD95143-5G BLDPBD95143-100MG BLDPBD95143-250MG
4-Bromo-6-methoxyquinoline 95%
4-Bromo-6-methoxyquinoline

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₈BrNO
MW: 238,0800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08437074
CAS číslo: 42881-66-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR