Tisk…

4-Chloro-2-iodophenol 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD95754-100GEA 13860 CZK
BLDPBD95754-0.025G BLDPBD95754-100G BLDPBD95754-1G BLDPBD95754-10G BLDPBD95754-250MG BLDPBD95754-5G BLDPBD95754-25G
4-Chloro-2-iodophenol 98%
4-Chloro-2-iodophenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₆H₄ClIO
MW: 254,4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235248
CAS číslo: 71643-66-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR