Tisk…

6-Amino-1-methyluracil 97%

BLDPBD9614-25GEA 456 CZK
BLDPBD9614-25G BLDPBD9614-100G BLDPBD9614-500G BLDPBD9614-5G BLDPBD9614-10G
6-Amino-1-methyluracil 97%
6-Amino-1-methyluracil

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₅H₇N₃O₂
MW: 141,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075366
CAS číslo: 2434-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR