3-(Hydroxymethyl)-5-methylisoxazole 98%
|
|
BLDPBD9615-250MGEA
684
CZK
BLDPBD9615-250MG
BLDPBD9615-5G
BLDPBD9615-25G
BLDPBD9615-1G
3-(Hydroxymethyl)-5-methylisoxazole 98%
3-(Hydroxymethyl)-5-methylisoxazole
Storage condition: Sealed in dry, 2-8°C
Vzorec:
C₅H₇NO₂ MW: 113,11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00085129 CAS číslo: 35166-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...