Tisk…

Thiophene-2-methylamine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD9621-1GEA 200 CZK
BLDPBD9621-1G BLDPBD9621-0.025G BLDPBD9621-5G BLDPBD9621-100G BLDPBD9621-10G BLDPBD9621-500G BLDPBD9621-25G
Thiophene-2-methylamine 97%
Thiophene-2-methylamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₅H₇NS
MW: 113,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005460
CAS číslo: 27757-85-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR