4-Fluoro-3-nitrobenzenesulphonamide 98%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD97219-10GEA
1830
CZK
BLDPBD97219-10G
BLDPBD97219-25G
BLDPBD97219-250MG
BLDPBD97219-100G
BLDPBD97219-5G
BLDPBD97219-1G
4-Fluoro-3-nitrobenzenesulphonamide 98%
4-Fluoro-3-nitrobenzenesulphonamide
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₆H₅FN₂O₄S MW: 220,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08703185 CAS číslo: 406233-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...