Tisk…

4-Fluorophenyl chloroformate 97%

BLDPBD97364-250MGEA 542 CZK
BLDPBD97364-250MG BLDPBD97364-1G
4-Fluorophenyl chloroformate 97%
4-Fluorophenyl chloroformate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₄ClFO₂
MW: 174,5569 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00013257
CAS číslo: 38377-38-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR