Tisk…

4-(Isopropoxycarbonyl)phenylboronic acid 98%

BLDPBD98212-1GEA 542 CZK
BLDPBD98212-1G BLDPBD98212-5G BLDPBD98212-25G BLDPBD98212-100G
4-(Isopropoxycarbonyl)phenylboronic acid 98%
4-(Isopropoxycarbonyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₃BO₄
MW: 208,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02259385
CAS číslo: 342002-82-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR