4-Isopropyl-2-nitroaniline 97%
|
|
BLDPBD98220-10GEA
15990
CZK
BLDPBD98220-10G
BLDPBD98220-25G
BLDPBD98220-5G
BLDPBD98220-250MG
BLDPBD98220-1G
4-Isopropyl-2-nitroaniline 97%
4-Isopropyl-2-nitroaniline
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₉H₁₂N₂O₂ MW: 180,204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00092920 CAS číslo: 63649-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...