Tisk…

3-Amino-4-methoxyphenylsulphonyl fluoride 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD98561-250MGEA 1570 CZK
BLDPBD98561-250MG BLDPBD98561-1G BLDPBD98561-100MG
3-Amino-4-methoxyphenylsulphonyl fluoride 95%
3-Amino-4-methoxyphenylsulphonyl fluoride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₈FNO₃S
MW: 205,2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007417
CAS číslo: 498-74-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR