Tisk…

4-Methoxypyridine-N-oxide hydrate 98%

BLDPBD98632-1GEA 285 CZK
BLDPBD98632-1G BLDPBD98632-5G BLDPBD98632-500G BLDPBD98632-25G BLDPBD98632-100G
4-Methoxypyridine-N-oxide hydrate 98%
4-Methoxypyridine-N-oxide hydrate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₇NO₂·H₂O
MW: 125,13 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149410
CAS číslo: 207511-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR