Tisk…

4-Methyl-3-oxopentanenitrile 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD98874-1GEA 656 CZK
BLDPBD98874-1G BLDPBD98874-10G BLDPBD98874-5G BLDPBD98874-250MG BLDPBD98874-25G
4-Methyl-3-oxopentanenitrile 97%
4-Methyl-3-oxopentanenitrile

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₆H₉NO
MW: 111,142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664144
CAS číslo: 29509-06-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR