Tisk…

4-Methyl-1H-pyrazole hydrochloride 95%

BLDPBD99116-1GEA 8410 CZK
BLDPBD99116-1G BLDPBD99116-100MG BLDPBD99116-250MG
4-Methyl-1H-pyrazole hydrochloride 95%
4-Methyl-1H-pyrazole hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₄H₇ClN₂
MW: 118,5648 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012707
CAS číslo: 56010-88-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR