Tisk…
VWR®, Sulphur in 20% biodiesel (B20) with Blank (2×100 ml)
Dodavatel: Avantor
VWR®
91704.001EA 3700 CZK
91704.001
VWR®, Sulphur in 20% biodiesel (B20) with Blank (2×100 ml)
Standardy Standardy kationické Standardy - síra
100 ml of each: Sulfur 0,0005% w/w in Biodiesel 20; Biodiesel (B20) Blank.
Upozornení: Toxic Substance.