Tisk…

VWR®, Sulphur in 20% biodiesel (B20) (0,0075% w/w), CRM

Dodavatel: Avantor
VWR®
91709.180EA 3030 CZK
91709.180
VWR®, Sulphur in 20% biodiesel (B20) (0,0075% w/w), CRM
Standardy Standardy kationické Standardy - síra
Certifikace: Certified reference material.

Upozornení: Carcinogen.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR