Tisk…

N(α)-Acetyl-D-tryptophan ≥98.0% (by HPLC, titration analysis)

Dodavatel: ABCR GMBH

ABCRAB171071.0001EA 0 CZK
ABCRAB171071.0001
N(α)-Acetyl-D-tryptophan ≥98.0% (by HPLC, titration analysis)
N(α)-Acetyl-D-tryptophan
Vzorec: C₁₃H₁₄N₂O₃
MW: 246,3 g/mol
MDL Number: MFCD00065021
CAS číslo: 2280-01-5
EINECS: 218-912-4
https://cz.vwr.com/stibo/web/std.lang.all/75/69/28927569.jpg

Objednat

Výsledky specifikačního testu

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -24.2 to -25.9 deg (C=2.5, methanol)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR