Tisk…
Experimentation kits, drosophila genotyping using PCR
Dodavatel: Edvotek
EDVO337EA 6840 CZK
EDVO337
Experimentation kits, drosophila genotyping using PCR
Materiály výukové Materiál pro výuku biologie Aktivity pro výuku PCR
Students learn about DNA polymorphisms by amplifying DNA regions that vary between wild and mutant Drosophila. Amplified DNA from wild-type and white-eyed flies are separated by agarose gel electrophoresis and analysed.
  • For 10 lab groups
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Thermal cycler, electrophoresis tank and power supply, automatic micropipette with tips, microwave, water bath, UV transilluminator and microcentrifuge.
Informace o dodávce: Kit includes instructions, PCR beads, primers, DNA extraction buffer, 200 bp ladder, proteinase K, Drosophila (wild-type and white-eye), agarose, electrophoresis buffer, and gel stain.