Tisk…
Experimentation kits, human PCR Tool Box™
Tool Box™
EDVO369EA 8010 CZK
EDVO369
Experimentation kits, human PCR Tool Box™
Polymerase Chain Reaction (PCR) is commonly used to determine paternity as it is a very sensitive method for DNA analysis. In this experiment, students gain an understanding of the principles behind this non-forensic use of DNA fingerprinting using their own DNA. The kit provides three sets of primers to carry out the PCR amplification of Alu element (PV92) on chromosome 16, the VNTR locus (D1S80) on chromosome 1, and two regions of the mitochondrial gene. Sufficient for six runs of each PCR reaction.
  • For 6 lab groups
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Informace o dodávce: Kit includes instructions, proteinase K, PCR beads, control and primer DNA, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.