Tisk…
Experimentation kits, electrophoretic properties of native proteins (agarose-based)
Dodavatel: Edvotek
EDVO111EA 4540 CZK
EDVO111
Experimentation kits, electrophoretic properties of native proteins (agarose-based)
Proteins are complex biomolecules with varying charge, size and shape that can be analysed by agarose gel electrophoresis. Gel analysis of native proteins enables students to evaluate natural charge and shape characteristics of proteins. Following electrophoresis, the protein samples are stained and visualised.
  • For 6 lab groups
  • Complete in one hour followed by overnight staining
Also required: Horizontal electrophoresis apparatus, power supply, white light visualisation system, 5 to 50 µl adjustable or 40 µl fixed volume micropipette, methanol, and glacial acetic acid.
Informace o dodávce: Kit includes instructions, protein samples, gel loading solution, agarose, electrophoresis buffer, and stain.