2-Thiouracil
Dodavatel: EHRENSTORFER GMBH
EHERC17561500EA
1520
CZK
EHERC17561500
2-Thiouracil
2-Thiouracil
Vzorec:
C₄H₄N₂OS MW: 128,15 g/mol Bod tání: 340 °C |
MDL Number:
MFCD00006039 CAS číslo: 141-90-2 EINECS: 205-508-8 Merck Index: 12,09504 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...