Tisk…

Methyl chloroacetate, Supelco®

Nebezpečí


M-Clarity™ quality level = MQ100

36546-1GEA 0 CZK
36546-1G
Methyl chloroacetate, Supelco®
Methyl chloroacetate
Vzorec: C₃H₅ClO₂
MW: 108,52 g/mol
Bod varu: 131 °C (1013 hPa)
Bod tání: –33 °C
Hustota: 1,2378 g/cm³ (20 °C)
Bod vzplanutí: 47 °C
MDL Number: MFCD00000931
CAS číslo: 96-34-4
EINECS: 202-501-1
UN: 2295
ADR: 6.1,I
Merck Index: 12,06122

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR