Tisk…

2-Bromethylamine hydrobromide

Varování

MOLE11263214-500GEA 0 CZK
MOLE11263214-500G
2-Bromethylamine hydrobromide
2-Bromethylamine hydrobromide
Vzorec: BrCH₂CH₂NH₃Br
MW: 204,89 g/mol
Bod tání: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS číslo: 2576-47-8
EINECS: 219-924-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR