Indole-4-carbaldehyd
MOLE61265833-1GEA
6040
CZK
MOLE61265833-1G
Indole-4-carbaldehyd
Indole-4-carbaldehyd
Vzorec:
C₉H₇NO MW: 145,16 g/mol |
MDL Number:
MFCD01632221 CAS číslo: 1074-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...