Tisk…

Trimethylsulphoxonium iodide

Dodavatel: Molekula

MOLEM82850263EA 0 CZK
MOLEM82850263
Trimethylsulphoxonium iodide
Trimethylsulphoxonium iodide
Vzorec: C₃H₉IOS
MW: 220,07 g/mol
Bod tání: 165…175 °C
MDL Number: MFCD00011899
CAS číslo: 1774-47-6
EINECS: 217-204-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR