Tisk…

Dimethylphenylphosphine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL265020-25GEA 24580 CZK
SIAL265020-25G SIAL265020-5G SIAL265020-1G
Dimethylphenylphosphine, Sigma-Aldrich®
Dimethylphenylphosphine
Vzorec: C₈H₁₁P
MW: 138,15 g/mol
Bod varu: 41…45 °C (0,1 mmHg)
Bod tání: 116…117 °C
Hustota: 0,9768 g/cm³ (11 °C)
MDL Number: MFCD00008509
CAS číslo: 672-66-2
EINECS: 211-595-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR