Tisk…

Allyl cyanoacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL360961-25GEA 4950 CZK
SIAL360961-25G
Allyl cyanoacetate, Sigma-Aldrich®
Allyl cyanoacetate
Vzorec: C₆H₇NO₂
MW: 125,13 g/mol
Bod tání: –40 °C
Hustota: 1,07 g/cm³ (20 °C)
Bod vzplanutí: 91 °C
MDL Number: MFCD00013817
CAS číslo: 13361-32-5
EINECS: 236-423-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR