Tisk…

N-Cyclohexylmaleimide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL381543-5GEA 4220 CZK
SIAL381543-5G
N-Cyclohexylmaleimide, Sigma-Aldrich®
N-Cyclohexylmaleimide
Vzorec: C₁₀H₁₃NO₂
MW: 179,22 g/mol
MDL Number: MFCD00043904
CAS číslo: 1631-25-0
EINECS: 216-630-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR