α-Bromo-3-nitro-p-toluic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL423564-5GEA
4240
CZK
SIAL423564-5G
α-Bromo-3-nitro-p-toluic acid, Sigma-Aldrich®
α-Bromo-3-nitro-p-toluic acid
Vzorec:
C₈H₆BrNO₄ MW: 260,04 g/mol Bod tání: 132 °C |
MDL Number:
MFCD00216575 CAS číslo: 55715-03-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...