Tisk…

2,5-Dibromothiazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL524182-5GEA 4680 CZK
SIAL524182-5G
2,5-Dibromothiazole, Sigma-Aldrich®
2,5-Dibromothiazole
Vzorec: C₃HBr₂NS
MW: 242,92 g/mol
MDL Number: MFCD00016891
CAS číslo: 4175-78-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR