4,6-Dihydroxypyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIALD120405-25GEA
0
CZK
SIALD120405-25G
4,6-Dihydroxypyrimidine, Sigma-Aldrich®
4,6-Dihydroxypyrimidine
Vzorec:
C₄H₄N₂O₂ MW: 112,09 g/mol Bod tání: > 300 °C |
MDL Number:
MFCD00016733 CAS číslo: 1193-24-4 EINECS: 214-772-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...