5-Fluoroindole-2-carboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIALF109-250MGEA
2270
CZK
SIALF109-250MG
5-Fluoroindole-2-carboxylic acid, Sigma-Aldrich®
5-Fluoroindole-2-carboxylic acid
Vzorec:
C₉H₆FNO₂ MW: 179,15 g/mol Bod tání: 258…259 °C |
MDL Number:
MFCD00005612 CAS číslo: 399-76-8 EINECS: 206-919-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...