Tisk…

o-Phenylenediamine dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ300

SIALP8787-100TABEA 11830 CZK
SIALP8787-100TAB
o-Phenylenediamine dihydrochloride, Sigma-Aldrich®
o-Phenylenediamine dihydrochloride
Vzorec: C₆H₁₀Cl₂N₂
MW: 181,06 g/mol
Bod tání: 205 °C
MDL Number: MFCD00012966
CAS číslo: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR