Tisk…
Cottonseed oil analytical standard, Supelco®
Dodavatel: Merck
47113.EA 0 CZK
47113.
Cottonseed oil analytical standard, Supelco®
Cottonseed oil
Hustota: 0,91 g/cm³ (20 °C)
MDL Number: MFCD00130872
CAS číslo: 8001-29-4
EINECS: 232-280-7